2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
5 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number FSA159240020
Price (MYR)
1,014.00
Add to cart
Quantity:
2 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,014.00
Add to cart
Chemical Identifiers
CAS99-20-7
IUPAC Name(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyHDTRYLNUVZCQOY-LBYRQFQKNA-N
SMILESOC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=98.5 %
Water=<1.0 % (K.F.)