Hydrocinnamic acid, 99%, Thermo Scientific Chemicals
Hydrocinnamic acid, 99%, Thermo Scientific Chemicals
Hydrocinnamic acid, 99%, Thermo Scientific Chemicals
Hydrocinnamic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrocinnamic acid, 99%, Thermo Scientific Chemicals
CAS: 501-52-0 | C9H10O2 | 150.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number FSA159101000
Price (MYR)
364.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
364.00
Add to cart
Chemical Identifiers
CAS27848-80-2
IUPAC Namelithium(1+) 2-hydroxypropanoate
Molecular FormulaC3H5LiO3
InChI KeyGKQWYZBANWAFMQ-UHFFFAOYNA-M
SMILES[Li+].CC(O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Specific optical rotation-13° to -16° (20°C, 589 nm) (c=1, H2O)
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration with HClO4>=98.5 %