Hydrocinnamic acid, 99%, Thermo Scientific Chemicals
Hydrocinnamic acid, 99%, Thermo Scientific Chemicals
Hydrocinnamic acid, 99%, Thermo Scientific Chemicals
Hydrocinnamic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrocinnamic acid, 99%, Thermo Scientific Chemicals
CAS: 501-52-0 | C9H10O2 | 150.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA159100050
Price (MYR)
252.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
252.00
Add to cart
Chemical Identifiers
CAS1344-28-1
IUPAC Namedialuminium(3+) trioxidandiide
Molecular FormulaAl2O3
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
CommentActivity: Brockman I
Particle SizeMean: ≥32µm to ≤63µm
FormPowder
pH≥4.0 to ≤5.0 (slurry, water, 5%)
Surface Area≥135 to ≤165 m²/g