Thiazolyl blue tetrazolium bromide, 98%, Thermo Scientific Chemicals
Thiazolyl blue tetrazolium bromide, 98%, Thermo Scientific Chemicals
Thiazolyl blue tetrazolium bromide, 98%, Thermo Scientific Chemicals
Thiazolyl blue tetrazolium bromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiazolyl blue tetrazolium bromide, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 mg
1 g
Packaging:
Glass bottle
Catalog number FSA158990050
Price (MYR)
2,226.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,226.00
Add to cart
Chemical Identifiers
CAS2991-42-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow or pale cream
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)≥97.5%
Melting Point (clear melt)28-34?C