Glycerol, 99+%, extra pure, Thermo Scientific Chemicals
Glycerol, 99+%, extra pure, Thermo Scientific Chemicals
Glycerol, 99+%, extra pure, Thermo Scientific Chemicals
Glycerol, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycerol, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 56-81-5 | C3H8O3 | 92.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
2.5 L
Packaging:
View all
Glass Bottle
Catalog number FSA158920010
Price (MYR)
490.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
490.00
Add to cart
Chemical Identifiers
CAS1537-52-6
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4400-1.4450 @ 20?C