Triphenylmethanol, 97%, Thermo Scientific Chemicals
Triphenylmethanol, 97%, Thermo Scientific Chemicals
Triphenylmethanol, 97%, Thermo Scientific Chemicals
Triphenylmethanol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triphenylmethanol, 97%, Thermo Scientific Chemicals
CAS: 76-84-6 | C19H16O | 260.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Plastic bottle
Catalog number FSA158910500
Price (MYR)
347.00
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
347.00
Add to cart
Chemical Identifiers
CAS2033-89-8
IUPAC Name3,4-dimethoxyphenol
Molecular FormulaC8H10O3
InChI KeySMFFZOQLHYIRDA-UHFFFAOYSA-N
SMILESCOC1=CC=C(O)C=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White to cream to brown or pale pink
CommentMay darken on storage
Melting Point (clear melt)72-82?C