Succinic Acid 99%, Thermo Scientific Chemicals
Succinic Acid 99%, Thermo Scientific Chemicals
Succinic Acid 99%, Thermo Scientific Chemicals
Succinic Acid 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Succinic Acid 99%, Thermo Scientific Chemicals
CAS: 110-15-6 | C4H6O4 | 118.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA158742500
Price (MYR)
255.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
255.00
Add to cart
Chemical Identifiers
CAS7787-61-3
IUPAC Namebismuth(3+) trifluoride
Molecular FormulaBiF3
InChI KeyBRCWHGIUHLWZBK-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Bi+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min