Succinic Acid 99%, Thermo Scientific Chemicals
Succinic Acid 99%, Thermo Scientific Chemicals
Succinic Acid 99%, Thermo Scientific Chemicals
Succinic Acid 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Succinic Acid 99%, Thermo Scientific Chemicals
CAS: 110-15-6 | C4H6O4 | 118.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
500 g
Packaging:
Plastic bottle
Catalog number FSA158740010
Price (MYR)
537.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
537.00
Add to cart
Chemical Identifiers
CAS19423-86-0
IUPAC Namethulium(3+) trichloride
Molecular FormulaCl3Tm
InChI KeyILOTUXNTERMOJL-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Tm+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.