trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals
trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals
trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals
trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals
CAS: 140-10-3 | C9H8O2 | 148.161 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Packaging:
View all
Plastic bottle
Catalog number FSA158571000
Price (MYR)
258.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
258.00
Add to cart
Chemical Identifiers
CAS622-47-9
IUPAC Name2-(4-methylphenyl)acetate
Molecular FormulaC9H9O2
InChI KeyGXXXUZIRGXYDFP-UHFFFAOYSA-M
SMILESCC1=CC=C(CC([O-])=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point90°C to 93°C
Appearance (Color)White
Infrared spectrumConforms
Titration with NaOH>=98.5 %