trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals
trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals
trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals
trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals
CAS: 140-10-3 | C9H8O2 | 148.161 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number FSA158570050
Price (MYR)
226.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
226.00
Add to cart
Chemical Identifiers
CAS142-25-6
IUPAC Name[2-(dimethylazaniumyl)ethyl](methyl)azanium
Molecular FormulaC5H16N2
InChI KeyHVOYZOQVDYHUPF-UHFFFAOYSA-P
SMILESC[NH2+]CC[NH+](C)C
View more
SpecificationsSpecification SheetSpecification Sheet
CommentPurchased in the U.S. and in other countries
FormLiquid
Assay from Suppliers CofA≥95.0% (GC, U.S. specification)
Refractive Index1.4180-1.4210 @ 20°C (non-U.S. specification)
Appearance (Color)Clear colorless to yellow