1,2,4-Trichlorobenzene, 99%, pure, Thermo Scientific Chemicals
1,2,4-Trichlorobenzene, 99%, pure, Thermo Scientific Chemicals
1,2,4-Trichlorobenzene, 99%, pure, Thermo Scientific Chemicals
1,2,4-Trichlorobenzene, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2,4-Trichlorobenzene, 99%, pure, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA157900025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤0.1%
Residue on Evaporation≤0.001%
Solution Testin water P.T.
Appearance (Color)Colorless
Color (APHA)≤10