Thymine, 99%, Thermo Scientific Chemicals
Thymine, 99%, Thermo Scientific Chemicals
Thymine, 99%, Thermo Scientific Chemicals
Thymine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thymine, 99%, Thermo Scientific Chemicals
CAS: 65-71-4 | C5H6N2O2 | 126.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number FSA157850250
Price (MYR)
369.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
369.00
Add to cart
Chemical Identifiers
CAS99-52-5
IUPAC Name2-methyl-4-nitroaniline
Molecular FormulaC7H8N2O2
InChI KeyXTTIQGSLJBWVIV-UHFFFAOYSA-N
SMILESCC1=CC(=CC=C1N)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or needles
Melting point129°C to 133°C
Appearance (Color)Yellow to brown to khaki
GC>=98.5 %