Indigo Carmine, High Purity Biological Stain
Indigo Carmine, High Purity Biological Stain
Indigo Carmine, High Purity Biological Stain
Indigo Carmine, High Purity Biological Stain
Thermo Scientific Chemicals
Indigo Carmine, High Purity Biological Stain
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA157445000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS106-24-1
IUPAC Name(2E)-3,7-dimethylocta-2,6-dien-1-ol
Molecular FormulaC10H18O
InChI KeyGLZPCOQZEFWAFX-JXMROGBWSA-N
SMILESCC(C)=CCC\C(C)=C\CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4750 to 1.4780 (20°C, 589 nm)
Indicator
RUO – Research Use Only