4-Nitrophenol, 99%, Thermo Scientific Chemicals
4-Nitrophenol, 99%, Thermo Scientific Chemicals
4-Nitrophenol, 99%, Thermo Scientific Chemicals
4-Nitrophenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitrophenol, 99%, Thermo Scientific Chemicals
CAS: 100-02-7 | C6H5NO3 | 139.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA157051000
Price (MYR)
328.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
328.00
Add to cart
Specifications
Chemical Name or Material4-Nitrophenol
CAS100-02-7
Health Hazard 1Warning
Melting Point112°C to 115°C
ColorYellow to Brown
View more
Indicator
RUO – Research Use Only