4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
CAS: 555-16-8 | C7H5NO3 | 151.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number FSA156995000
Price (MYR)
2,755.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
2,755.00
Add to cart
Chemical Identifiers
CAS2409-55-4
IUPAC Name2-tert-butyl-4-methylphenol
Molecular FormulaC11H16O
InChI KeyIKEHOXWJQXIQAG-UHFFFAOYSA-N
SMILESCC1=CC=C(O)C(=C1)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
FormFused solid
Appearance (Color)White to pale cream
Identification (FTIR)Conforms
Assay (GC)≥98.5%
Melting Point (clear melt)49.0-55.0?C