4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
CAS: 555-16-8 | C7H5NO3 | 151.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number FSA156990250
Price (MYR)
364.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
364.00
Add to cart
Chemical Identifiers
CAS556-90-1
IUPAC Name2-amino-4,5-dihydro-1,3-thiazol-4-one
Molecular FormulaC3H4N2OS
InChI KeyHYMJHROUVPWYNQ-UHFFFAOYSA-N
SMILESNC1=NC(=O)CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%