Isovaleric acid, 99%, Thermo Scientific Chemicals
Isovaleric acid, 99%, Thermo Scientific Chemicals
Isovaleric acid, 99%, Thermo Scientific Chemicals
Isovaleric acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isovaleric acid, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
10 g
100 g
Packaging:
Glass bottle
Catalog number FSA156695000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS118-29-6
IUPAC Name2-(hydroxymethyl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC9H7NO3
InChI KeyMNSGOOCAMMSKGI-UHFFFAOYSA-N
SMILESOCN1C(=O)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystalline powder
Identification (FTIR)Conforms
Melting Point (clear melt)138-148?C
Assay (HPLC)≥96.0%