3,5-Dinitrosalicylic acid, 98%, Thermo Scientific Chemicals
3,5-Dinitrosalicylic acid, 98%, Thermo Scientific Chemicals
3,5-Dinitrosalicylic acid, 98%, Thermo Scientific Chemicals
3,5-Dinitrosalicylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,5-Dinitrosalicylic acid, 98%, Thermo Scientific Chemicals
3,5-dinitrosalicylic acid, CAS # 609-99-4, also known as DNS or DNSA, is used for quantifying carbohydrate levels in blood by calorimetry.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA156440010
Price (MYR)
4,124.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
4,124.00
Add to cart
Chemical Identifiers
CAS7439-89-6
IUPAC Nameiron
Molecular FormulaFe
InChI KeyXEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES[Fe]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 0.25 mm (0.01 in.) diameter
Total Metal Impurities≤0.01%
General Description
3,5-Dinitrosalicylic acid also known as DNA or DNSA reacts with reducing sugars forming 3-amino-5-nitrosalicylic acid, which is an aromatic compound with maximum absorption at 540 nm allowing spectrophotometer measurement of the amount of reducing molecules
Applications
  • It is used in the quantification of reducing sugars by colorimetry in the blood. It is also used to analyze glycosidase (glycoside hydrolase) activity by quantitation of enzymatically released reducing sugar
  • 3,5-Dinitrosalicylic acid can detect reducing substances in the urine
  • This compound is used to detect the presence of free carbonyl groups (C=O)
  • 3,5-Dinitrosalicylic acid can be used to determine the effects of silver nanoparticles (AgNPs) on membrane leakage
RUO – Research Use Only