2,2,4,4,6,8,8-Heptamethylnonane, 98%, Thermo Scientific Chemicals
2,2,4,4,6,8,8-Heptamethylnonane, 98%, Thermo Scientific Chemicals
2,2,4,4,6,8,8-Heptamethylnonane, 98%, Thermo Scientific Chemicals
2,2,4,4,6,8,8-Heptamethylnonane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2,4,4,6,8,8-Heptamethylnonane, 98%, Thermo Scientific Chemicals
CAS: 4390-04-9 | C16H34 | 226.44 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA1562500101 LGlass Bottle
Catalog number FSA156250010
Price (MYR)
5,531.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
5,531.00
EA
Add to cart
Specifications
Chemical Name or Material2,2,4,4,6,8,8-Heptamethylnonane
CAS4390-04-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0