L(+)-Glutamic acid, 99%
L(+)-Glutamic acid, 99%
L(+)-Glutamic acid, 99%
L(+)-Glutamic acid, 99%
Thermo Scientific Chemicals
L(+)-Glutamic acid, 99%
CAS: 56-86-0 | C5H9NO4 | 147.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
1 kg
Packaging:
Plastic Bottle
Catalog number FSA156212500
Price (MYR)
278.00
Add to cart
Quantity:
250 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
278.00
Add to cart
Specifications
Chemical Name or MaterialL(+)-Glutamic acid
Ammonium200ppm max.
CAS56-86-0
ColorWhite
Chlorine (Cl)200ppm max.