Diphenylborinic acid 2-aminoethyl ester, 98%, Thermo Scientific Chemicals
Diphenylborinic acid 2-aminoethyl ester, 98%, Thermo Scientific Chemicals
Diphenylborinic acid 2-aminoethyl ester, 98%, Thermo Scientific Chemicals
Diphenylborinic acid 2-aminoethyl ester, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diphenylborinic acid 2-aminoethyl ester, 98%, Thermo Scientific Chemicals
CAS: 524-95-8 | C14H16BNO | 225.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
10 g
Packaging:
Glass bottle
Catalog number FSA155420250
Price (MYR)
1,617.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,617.00
Add to cart
Chemical Identifiers
CAS303-98-0
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=97.5 %
Specific absorbance A (1%/1cm)>=162 (275 nm, EtOH)
Appearance (Color)Yellow to orange
Appearance (Form)Crystalline powder