2'-Deoxyadenosine hydrate, 99%
2'-Deoxyadenosine hydrate, 99%
2'-Deoxyadenosine hydrate, 99%
2'-Deoxyadenosine hydrate, 99%
Thermo Scientific Chemicals
2'-Deoxyadenosine hydrate, 99%
CAS: 16373-93-6 | C10H13N5O3 | 251.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA154990250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10025-82-8
IUPAC Nameindium(3+) trichloride
Molecular FormulaCl3In
InChI KeyPSCMQHVBLHHWTO-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[In+3]
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Appearance (Color)White
Chloride content≥47.12%
Assay from Suppliers CofA≥98.0%