Phthalic Anhydride, 99%, Thermo Scientific Chemicals
Phthalic Anhydride, 99%, Thermo Scientific Chemicals
Phthalic Anhydride, 99%, Thermo Scientific Chemicals
Phthalic Anhydride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phthalic Anhydride, 99%, Thermo Scientific Chemicals
CAS: 85-44-9 | C8H4O3 | 148.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA154960025
Price (MYR)
510.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
510.00
Add to cart
Chemical Identifiers
CAS556-08-1
IUPAC Name4-acetamidobenzoic acid
Molecular FormulaC9H9NO3
InChI KeyQCXJEYYXVJIFCE-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=99.0 %
Appearance (Color)Off-white to light beige
Infrared spectrumConforms
Titration with NaOH99.0 to 101.0 %
Appearance (Form)Powder