Phthalic Anhydride, 99%, Thermo Scientific Chemicals
Phthalic Anhydride, 99%, Thermo Scientific Chemicals
Phthalic Anhydride, 99%, Thermo Scientific Chemicals
Phthalic Anhydride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phthalic Anhydride, 99%, Thermo Scientific Chemicals
CAS: 85-44-9 | C8H4O3 | 148.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA154960010
Price (MYR)
321.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
321.00
Add to cart
Chemical Identifiers
CAS7722-84-1
IUPAC Nameperoxol
Molecular FormulaH2O2
InChI KeyMHAJPDPJQMAIIY-UHFFFAOYSA-N
SMILESOO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear solution
Heavy metals=<10 ppm
Chloride (Cl)=<2 ppm
Stabilizerabout 0.03% stabilizers
Color scale=<10 APHA