Citraconic anhydride, 98%, Thermo Scientific Chemicals
Citraconic anhydride, 98%, Thermo Scientific Chemicals
Citraconic anhydride, 98%, Thermo Scientific Chemicals
Citraconic anhydride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citraconic anhydride, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
Packaging:
Glass bottle
Catalog number FSA154791000
Price (MYR)
460.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
460.00
Add to cart
Chemical Identifiers
CAS15164-44-0
IUPAC Name4-iodobenzaldehyde
Molecular FormulaC7H5IO
InChI KeyNIEBHDXUIJSHSL-UHFFFAOYSA-N
SMILESIC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Melting point75°C to 80°C
Appearance (Color)White to yellow to beige
Infrared spectrumConforms
HPLC>=97.0 %