4-Carboxybenzaldehyde, 96%, Thermo Scientific Chemicals
4-Carboxybenzaldehyde, 96%, Thermo Scientific Chemicals
4-Carboxybenzaldehyde, 96%, Thermo Scientific Chemicals
4-Carboxybenzaldehyde, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Carboxybenzaldehyde, 96%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA154580250
Price (MYR)
643.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
643.00
Add to cart
Chemical Identifiers
CAS131-17-9
IUPAC Name1,2-bis(prop-2-en-1-yl) benzene-1,2-dicarboxylate
Molecular FormulaC14H14O4
InChI KeyQUDWYFHPNIMBFC-UHFFFAOYSA-N
SMILESC=CCOC(=O)C1=CC=CC=C1C(=O)OCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Refractive index1.5180 to 1.5200 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=97.5 %