DL-Camphoroquinone, 99%, Thermo Scientific Chemicals
DL-Camphoroquinone, 99%, Thermo Scientific Chemicals
DL-Camphoroquinone, 99%, Thermo Scientific Chemicals
DL-Camphoroquinone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Camphoroquinone, 99%, Thermo Scientific Chemicals
CAS: 10373-78-1 | C10H14O2 | 166.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
Packaging:
Glass bottle
Catalog number FSA154570500
Price (MYR)
2,341.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,341.00
Add to cart
Chemical Identifiers
CAS606-23-5
IUPAC Name2,3-dihydro-1H-indene-1,3-dione
Molecular FormulaC9H6O2
InChI KeyUHKAJLSKXBADFT-UHFFFAOYSA-N
SMILESO=C1CC(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to beige to golden
Infrared spectrumConforms
Melting point124°C to 131°C
Titration with NaOH>=96.0 %
Appearance (Form)Powder