4-tert-Butylcatechol, 99%, Thermo Scientific Chemicals
4-tert-Butylcatechol, 99%, Thermo Scientific Chemicals
4-tert-Butylcatechol, 99%, Thermo Scientific Chemicals
4-tert-Butylcatechol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-tert-Butylcatechol, 99%, Thermo Scientific Chemicals
CAS: 98-29-3 | C10H14O2 | 166.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
View all
Plastic Bottle
Catalog number FSA154350025
Price (MYR)
1,651.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
1,651.00
Add to cart
Chemical Identifiers
CAS1308-38-9
IUPAC Namedichromium(3+) trioxidandiide
Molecular FormulaCr2O3
InChI KeyUOUJSJZBMCDAEU-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Cr+3].[Cr+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.03% max.