2-Bromothiophene, 98%, Thermo Scientific Chemicals
2-Bromothiophene, 98%, Thermo Scientific Chemicals
2-Bromothiophene, 98%, Thermo Scientific Chemicals
2-Bromothiophene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromothiophene, 98%, Thermo Scientific Chemicals
CAS: 1003-09-4 | C4H3BrS | 163.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
Packaging:
Glass bottle
Catalog number FSA154290500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS110-15-6
IUPAC Namebutanedioic acid
Molecular FormulaC4H6O4
InChI KeyKDYFGRWQOYBRFD-UHFFFAOYSA-N
SMILESOC(=O)CCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=98.5 %
Fumaric and maleic acid=<1 %
Appearance (Form)Crystalline powder
Melting point185°C to 190°C
Appearance (Color)White