Benzenesulfinic acid, sodium salt, 97%, Thermo Scientific Chemicals
Benzenesulfinic acid, sodium salt, 97%, Thermo Scientific Chemicals
Benzenesulfinic acid, sodium salt, 97%, Thermo Scientific Chemicals
Benzenesulfinic acid, sodium salt, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzenesulfinic acid, sodium salt, 97%, Thermo Scientific Chemicals
CAS: 873-55-2 | C6H5NaO2S | 164.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number FSA154051000
Price (MYR)
438.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
438.00
Add to cart
Chemical Identifiers
CAS139631-62-2
IUPAC Namecyclopropanesulfonyl chloride
Molecular FormulaC3H5ClO2S
InChI KeyPFWWSGFPICCWGU-UHFFFAOYSA-N
SMILESClS(=O)(=O)C1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
FormLiquid
Refractive Index1.4740-1.4790 @ 20?C