3-Aminophthalhydrazide, 98%, pure, Thermo Scientific Chemicals
3-Aminophthalhydrazide, 98%, pure, Thermo Scientific Chemicals
3-Aminophthalhydrazide, 98%, pure, Thermo Scientific Chemicals
3-Aminophthalhydrazide, 98%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Aminophthalhydrazide, 98%, pure, Thermo Scientific Chemicals
3-Aminophthalhydrazide, 98%, pure, Acros Organics, Quantity: 25g, Packaging: Glass bottle, Blue-Purple to Green, Grade: Pure, Melting Point: 319.0 deg.C to 320.0 deg.C, Molecular Weight: 177.16, Percent Purity: 98%, Assay Percent Range: 98%, Beilstein: 25,IV,4192, CAS: 521-31-3, CAS Max %: 100.0
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number FSA153850050
Price (MYR)
459.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
459.00
Add to cart
Chemical Identifiers
CAS5593-70-4
IUPAC Nametitanium(4+) tetrakis(butan-1-olate)
Molecular FormulaC16H36O4Ti
InChI KeyYHWCPXVTRSHPNY-UHFFFAOYSA-N
SMILES[Ti+4].CCCC[O-].CCCC[O-].CCCC[O-].CCCC[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to pale brown
Chloride content≤50 ppm (U.S. specification)
Gravimetric analysisAs titanium dioxide, TiO2: 23.0-23.9% (non-U.S. specification)
Viscosity50-90cP @ 25°C (non-U.S. specification)