4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
25 g
500 g
Packaging:
View all
Plastic bottle
Catalog number FSA153801000
Price (MYR)
2,441.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
2,441.00
Add to cart
Specifications
Chemical Name or Material4-Aminohippuric acid
CAS61-78-9
CAS Min %98.5
Health Hazard 1Warning
CAS Max %100.0