4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Aminohippuric acid, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number FSA153800100
Price (MYR)
462.00
Add to cart
Quantity:
10 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
462.00
Add to cart
Chemical Identifiers
CAS14804-31-0
IUPAC Name4-bromo-1-methoxy-2-methylbenzene
Molecular FormulaC8H9BrO
InChI KeyUDLRGQOHGYWLCS-UHFFFAOYSA-N
SMILESCOC1=C(C)C=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Melting Point (clear melt)64.5-70.5°C (non-U.S. specification)