Ethyl levulinate, 98%, Thermo Scientific Chemicals
Ethyl levulinate, 98%, Thermo Scientific Chemicals
Ethyl levulinate, 98%, Thermo Scientific Chemicals
Ethyl levulinate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl levulinate, 98%, Thermo Scientific Chemicals
CAS: 539-88-8 | C7H12O3 | 144.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA153625000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5452-35-7
IUPAC Namecycloheptanamine
Molecular FormulaC7H15N
InChI KeyVXVVUHQULXCUPF-UHFFFAOYSA-N
SMILESNC1CCCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Refractive index1.4710 to 1.4730 (20°C, 589 nm)
Water=<0.5 % (K.F.)
Appearance (Color)Clear colorless to light yellow