2-Deoxy-D-ribose, 99%
2-Deoxy-D-ribose, 99%
2-Deoxy-D-ribose, 99%
2-Deoxy-D-ribose, 99%
Thermo Scientific Chemicals
2-Deoxy-D-ribose, 99%
CAS: 533-67-5 | C5H10O4 | 134.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA153280050
Price (MYR)
540.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
540.00
Add to cart
Chemical Identifiers
CAS63148-62-9
IUPAC NamePolydimethylsiloxane
Molecular Formula(C2H6OSi)n
SMILESC[Si](C)(-*)O-*
MDL NumberMFCD00132673
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4030 to 1.4060 (20°C, 589 nm)
Appearance (Form)Oily liquid
Viscosity475 to 525 mm²/s (at 25°C)