Giemsa Stain, High Purity Biological Stain
Giemsa Stain, High Purity Biological Stain
Giemsa Stain, High Purity Biological Stain
Giemsa Stain, High Purity Biological Stain
Thermo Scientific Chemicals
Giemsa Stain, High Purity Biological Stain
CAS: 51811-82-6 | C14H14ClN3S | 291.80 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA153230250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Absorbance950 min. (in MeOH, on dried substance), 600 min. (in MeOH, on dried substance) A (1%/1cm)
Chemical Name or MaterialGiemsa Stain
Melting Point300.0°C
CAS51811-82-6
EINECS Number257-438-2