o-Vanillin, 99%, Thermo Scientific Chemicals
o-Vanillin, 99%, Thermo Scientific Chemicals
o-Vanillin, 99%, Thermo Scientific Chemicals
o-Vanillin, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
o-Vanillin, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
Packaging:
Glass bottle
Catalog number FSA153071000
Price (MYR)
480.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
480.00
Add to cart
Chemical Identifiers
CAS614-18-6
IUPAC Nameethyl pyridine-3-carboxylate
Molecular FormulaC8H9NO2
InChI KeyXBLVHTDFJBKJLG-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5015-1.5045 @ 20*C
Appearance (Color)Clear colorless to pale yellow or pale orange
Assay (GC)≥98.5%