Amaranth, 85%
Amaranth, 85%
Amaranth, 85%
Thermo Scientific Chemicals
Amaranth, 85%
CAS: 915-67-3 | C20H11N2Na3O10S3 | 604.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA153035000
Price (MYR)
2,298.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,298.00
Add to cart
Chemical Identifiers
CAS7782-89-0
IUPAC Namelithium(1+) azanide
Molecular FormulaH2LiN
InChI KeyAFRJJFRNGGLMDW-UHFFFAOYSA-N
SMILES[Li+].[NH2-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey
FormPowder (may contain black specks)
Assay (unspecified)≥94.0%