Naphthol Green B
Naphthol Green B
Naphthol Green B
Thermo Scientific Chemicals
Naphthol Green B
CAS: 19381-50-1 | C30H18FeN3Na3O15S3 | 881.474 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA152931000
Price (MYR)
356.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
356.00
Add to cart
Chemical Identifiers
CAS7784-24-9
IUPAC Namealuminium(3+) potassium dodecahydrate disulfate
Molecular FormulaAlH24KO20S2
InChI KeyGNHOJBNSNUXZQA-UHFFFAOYSA-J
SMILESO.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystals
Titration Complexometric98.0 to 102.0 %
Heavy metals (as Pb)=<10 ppm
Insoluble matter=<0.005 %
View more
Indicator
RUO – Research Use Only