Eosin Y, pure, high purity, biological stain
Eosin Y, pure, high purity, biological stain
Eosin Y, pure, high purity, biological stain
Eosin Y, pure, high purity, biological stain
Thermo Scientific Chemicals
Eosin Y, pure, high purity, biological stain
CAS: 17372-87-1 | C20H12Br4Na2O8 | 745.904 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA152881000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS110-94-1
IUPAC Namepentanedioic acid
Molecular FormulaC5H8O4
InChI KeyJFCQEDHGNNZCLN-UHFFFAOYSA-N
SMILESOC(=O)CCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.5%
Appearance (Color)White to pale cream
Assay (Silylated GC)≥98.5%
Melting Point95.0-101.0?C
FormCrystals or powder or crystalline powder
View more
Indicator
RUO – Research Use Only