1-Bromopentane, 98%, Thermo Scientific Chemicals
1-Bromopentane, 98%, Thermo Scientific Chemicals
1-Bromopentane, 98%, Thermo Scientific Chemicals
1-Bromopentane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromopentane, 98%, Thermo Scientific Chemicals
CAS: 110-53-2 | C5H11Br | 151.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA152522500
Price (MYR)
501.00
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
501.00
Add to cart
Chemical Identifiers
CAS10060-12-5
IUPAC Namechromium(3+) hexahydrate trichloride
Molecular FormulaCl3CrH12O6
InChI KeyLJAOOBNHPFKCDR-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or crystalline powder
Assay from Supplier's CofA≥97.0%
Appearance (Color)Dark green
Impurity content≤0.01% Iron (Fe)
Impurity content≤0.01% Sulfate (SO₄)