1-Bromopentane, 98%, Thermo Scientific Chemicals
1-Bromopentane, 98%, Thermo Scientific Chemicals
1-Bromopentane, 98%, Thermo Scientific Chemicals
1-Bromopentane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromopentane, 98%, Thermo Scientific Chemicals
CAS: 110-53-2 | C5H11Br | 151.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Glass bottle
Catalog number FSA152520010
Price (MYR)
1,575.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,575.00
Add to cart
Chemical Identifiers
CAS10294-54-9
IUPAC Namedicaesium(1+) sulfate
Molecular FormulaCs2O4S
InChI KeyFLJPGEWQYJVDPF-UHFFFAOYSA-L
SMILES[Cs+].[Cs+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.