1-Adamantanecarboxylic acid chloride, 97%, Thermo Scientific Chemicals
1-Adamantanecarboxylic acid chloride, 97%, Thermo Scientific Chemicals
1-Adamantanecarboxylic acid chloride, 97%, Thermo Scientific Chemicals
1-Adamantanecarboxylic acid chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Adamantanecarboxylic acid chloride, 97%, Thermo Scientific Chemicals
CAS: 2094-72-6 | C11H15ClO | 198.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA152480250
Price (MYR)
1,148.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,148.00
Add to cart
Chemical Identifiers
CAS30389-18-5
IUPAC Name1-ethynylcyclohexan-1-amine
Molecular FormulaC8H13N
InChI KeyGDKOYYDQISQOMH-UHFFFAOYSA-N
SMILESNC1(CCCCC1)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to pale brown
Assay (GC)>97.5%