p-Anisic acid, 98%, Thermo Scientific Chemicals
p-Anisic acid, 98%, Thermo Scientific Chemicals
p-Anisic acid, 98%, Thermo Scientific Chemicals
p-Anisic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Anisic acid, 98%, Thermo Scientific Chemicals
CAS: 100-09-4 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
Packaging:
Plastic bottle
Catalog number FSA152371000
Price (MYR)
279.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
279.00
Add to cart
Chemical Identifiers
CAS541-73-1
IUPAC Name1,3-dichlorobenzene
Molecular FormulaC6H4Cl2
InChI KeyZPQOPVIELGIULI-UHFFFAOYSA-N
SMILESClC1=CC(Cl)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Water=<0.1 %