Trichloroacetic acid, 99%, extra pure, Thermo Scientific Chemicals
Trichloroacetic acid, 99%, extra pure, Thermo Scientific Chemicals
Trichloroacetic acid, 99%, extra pure, Thermo Scientific Chemicals
Trichloroacetic acid, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trichloroacetic acid, 99%, extra pure, Thermo Scientific Chemicals
CAS: 76-03-9 | C2HCl3O2 | 163.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Packaging:
Plastic bottle
Catalog number FSA152130025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTrichloroacetic acid
Name NoteExtra Pure
CAS76-03-9
Melting Point52.0°C to 58.0°C
Boiling Point196.0°C