2,2,6,6-Tetramethylpiperidine, 98%, Thermo Scientific Chemicals
2,2,6,6-Tetramethylpiperidine, 98%, Thermo Scientific Chemicals
2,2,6,6-Tetramethylpiperidine, 98%, Thermo Scientific Chemicals
2,2,6,6-Tetramethylpiperidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2,6,6-Tetramethylpiperidine, 98%, Thermo Scientific Chemicals
CAS: 768-66-1 | C9H19N | 141.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
25 g
Packaging:
Glass bottle
Catalog number FSA151951000
Price (MYR)
1,704.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,704.00
Add to cart
Chemical Identifiers
CAS5177-27-5
IUPAC Name2,4-dichloropyrimidin-5-amine
Molecular FormulaC4H3Cl2N3
InChI KeyRINHVELYMZLXIW-UHFFFAOYSA-N
SMILESNC1=CN=C(Cl)N=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to pale yellow
Melting Point (clear melt)116.0-126.0?C
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%