2-Bromo-3'-methoxyacetophenone, 97%, Thermo Scientific Chemicals
2-Bromo-3'-methoxyacetophenone, 97%, Thermo Scientific Chemicals
2-Bromo-3'-methoxyacetophenone, 97%, Thermo Scientific Chemicals
2-Bromo-3'-methoxyacetophenone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromo-3'-methoxyacetophenone, 97%, Thermo Scientific Chemicals
CAS: 5000-65-7 | C9H9BrO2 | 229.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
Packaging:
Glass bottle
Catalog number FSA151910250
Price (MYR)
847.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
847.00
Add to cart
Chemical Identifiers
CAS7789-78-8
IUPAC Namecalcium
Molecular FormulaCa
InChI KeyOYPRJOBELJOOCE-UHFFFAOYSA-N
SMILES[Ca]
View more
SpecificationsSpecification SheetSpecification Sheet
Aluminium (Al)=<100 ppm
Appearance (Color)Grey
Assay>=92 % (Total Ca)
Iron (Fe)=<100 ppm
Particle size=<2 mm