2-Bromo-3'-methoxyacetophenone, 97%, Thermo Scientific Chemicals
2-Bromo-3'-methoxyacetophenone, 97%, Thermo Scientific Chemicals
2-Bromo-3'-methoxyacetophenone, 97%, Thermo Scientific Chemicals
2-Bromo-3'-methoxyacetophenone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromo-3'-methoxyacetophenone, 97%, Thermo Scientific Chemicals
CAS: 5000-65-7 | C9H9BrO2 | 229.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Packaging:
Glass bottle
Catalog number FSA151910100
Price (MYR)
503.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
503.00
Add to cart
Chemical Identifiers
CAS140-67-0
IUPAC Name1-methoxy-4-(prop-2-en-1-yl)benzene
Molecular FormulaC10H12O
InChI KeyZFMSMUAANRJZFM-UHFFFAOYSA-N
SMILESCOC1=CC=C(CC=C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5195-1.5235 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥97.5%