1-Adamantanamine hydrochloride, 99+%, Thermo Scientific Chemicals
1-Adamantanamine hydrochloride, 99+%, Thermo Scientific Chemicals
1-Adamantanamine hydrochloride, 99+%, Thermo Scientific Chemicals
1-Adamantanamine hydrochloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Adamantanamine hydrochloride, 99+%, Thermo Scientific Chemicals
CAS: 665-66-7 | C10H18ClN | 187.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number FSA151670250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS7778-77-0
IUPAC Namepotassium dihydrogen phosphate
Molecular FormulaH2KO4P
InChI KeyGNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES[K+].OP(O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
UVat 280 nm A: =<0.005
Appearance (Color)Colorless to white
UV(0.1 M in water)
Heavy metals (as Pb)=<0.001 %
View more
Applications
1-Adamantanamine hydrochloride is used in the prophylactic or symptomatic treatment of influenza A. It is also used as an antiparkinsonian agent, to treat extra pyramidal reactions, and for postherpetic neuralgia. It is also used an NMDA-receptor antagoinst.
Solubility
Soluble in water (50 mg/ml), ethanol (38 mg/ml at 25°C), chloroform (1:18), DMSO (38 mg/ml at 25°C), and methanol
Notes
Hygroscopic. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent and water/moisture.
RUO – Research Use Only