Myricetin, 95%
Myricetin, 95%
Myricetin, 95%
Thermo Scientific Chemicals
Myricetin, 95%
CAS: 529-44-2 | C15H10O8 | 318.24 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA15159025025 mgGlass bottle
Catalog number FSA151590250
Price (MYR)
539.00
Add to cart
Quantity:
25 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
539.00
Add to cart
Specifications
Chemical Name or MaterialMyricetin
Name Note95%
CAS529-44-2
CAS Min %94.0
CAS Max %100.0